Anti-EDN1 Mouse Monoclonal Antibody [clone: 10C85]

Lieferant: US Biological
E3098-24B
USBIE3098-24BEA 506 EUR
USBIE3098-24B
Anti-EDN1 Mouse Monoclonal Antibody [clone: 10C85]
Antikörper
Type: Primary
Antigen: EDN1 (endothelin 1)
Clonality: monoclonal
Clone: 10C85
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR