Anti-SLC29A1 Rabbit Polyclonal Antibody

Lieferant: US Biological
E3225-01
USBIE3225-01EA 649 EUR
USBIE3225-01
Anti-SLC29A1 Rabbit Polyclonal Antibody
Antikörper
  • Recommended dilution: Western Blot: 1:1000

Type: Primary
Antigen: SLC29A1 (solute carrier family 29 (equilibrative nucleoside transporter), member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR