Anti-Enterococcus sp. Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
E3270-01B
USBIE3270-01BEA 459 EUR
USBIE3270-01B
Anti-Enterococcus sp. Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: Enterococcus sp.
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR