Anti-EOMES Sheep polyclonal antibody

Lieferant: US Biological
E3320-01
USBIE3320-01EA 897 EUR
USBIE3320-01
Anti-EOMES Sheep polyclonal antibody
Antikörper
Recommended dilution: Western Blot: 1 ?g/ml

Type: Primary
Antigen: EOMES (eomesodermin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR