Anti-EOMES Mouse monoclonal antibody [clone: 11n64]

Lieferant: US Biological
E3320-01C
USBIE3320-01CEA 897 EUR
USBIE3320-01C
Anti-EOMES Mouse monoclonal antibody [clone: 11n64]
Antikörper
Recommended dilution: Western Blot: 1 ?g/ml; Immunochemistry: 8-25 ?g/ml;

Type: Primary
Antigen: EOMES (eomesodermin)
Clonality: monoclonal
Clone: 11n64
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR