Anti-EOMES Chicken polyclonal antibody

Lieferant: US Biological
E3321
USBIE3321EA 817 EUR
USBIE3321
Anti-EOMES Chicken polyclonal antibody
Antikörper
Recommended dilution: Western Blot: 0. 05 ?g/ml

Type: Primary
Antigen: EOMES (eomesodermin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR