Anti-Epoxide Hydrolase 2, Soluble Rabbit Polyclonal Antibody

Lieferant: US Biological
E3429-07
USBIE3429-07EA 681 EUR
USBIE3429-07
Anti-Epoxide Hydrolase 2, Soluble Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Immunochemistry: 20 ?g/ml

Type: Primary
Antigen: Epoxide Hydrolase 2, Soluble
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR