Anti-ERBB2 Rabbit Polyclonal Antibody
Lieferant: US Biological
E3451-27Q
USBIE3451-27QEA 610 EUR
USBIE3451-27Q
Anti-ERBB2 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:5000-1:20000; Immunochemistry: 1:50-1:200
Type: Primary
Antigen: ERBB2 (v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Zebrafish