Anti-ERG Goat Polyclonal Antibody

Lieferant: US Biological
E3452-01D
USBIE3452-01DEA 632 EUR
USBIE3452-01D
Anti-ERG Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:16000; Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: ERG (v-ets avian erythroblastosis virus E26 oncogene homolog)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR