Anti-ERG Goat Polyclonal Antibody
Lieferant: US Biological
E3452-01E
USBIE3452-01EEA 640 EUR
USBIE3452-01E
Anti-ERG Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:32000; Immunochemistry: 4-8?g/ml
Type: Primary
Antigen: ERG (v-ets avian erythroblastosis virus E26 oncogene homolog)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat