Anti-Estriol Mouse Monoclonal Antibody [clone: 9F13]

Lieferant: US Biological
E3560-02F
USBIE3560-02FEA 523 EUR
USBIE3560-02F
Anti-Estriol Mouse Monoclonal Antibody [clone: 9F13]
Antikörper
Type: Primary
Antigen: Estriol
Clonality: monoclonal
Clone: 9F13
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR