Anti-Estriol Sheep Polyclonal Antibody
Lieferant: US Biological
E3560-02K
USBIE3560-02KEA 473 EUR
USBIE3560-02K
Anti-Estriol Sheep Polyclonal Antibody
Antikörper
Recommended dilution: Radio Immunoassay: 1:30000
Type: Primary
Antigen: Estriol
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity:
Bestellen