Anti-ESR1 Rabbit Polyclonal Antibody
Lieferant: US Biological
E3565-10K
USBIE3565-10KEA 489 EUR
USBIE3565-10K
Anti-ESR1 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Immunochemistry: 1:50-1:100
Type: Primary
Antigen: ESR1 (estrogen receptor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human