Anti-Estrone Glucuronide Sheep Polyclonal Antibody

Lieferant: US Biological
E3570-50B
USBIE3570-50BEA 394 EUR
USBIE3570-50B
Anti-Estrone Glucuronide Sheep Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Estrone Glucuronide
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR