Anti-EIF3M Goat polyclonal antibody

Lieferant: US Biological
E8949-37T1
USBIE8949-37T1EA 640 EUR
USBIE8949-37T1
Anti-EIF3M Goat polyclonal antibody
Antikörper
Recommended dilution: ELISA: 1:32000; Western Blot: 1-3?g/ml

Type: Primary
Antigen: EIF3M (eukaryotic translation initiation factor 3, subunit M)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Dog, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR