Anti-Factor VIII, Human Sheep Polyclonal Antibody
Lieferant: US Biological
F0016-10-1
USBIF0016-10-1EA 466 EUR
USBIF0016-10-1
Anti-Factor VIII, Human Sheep Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Factor VIII, Human
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human