Anti-F9 Mouse monoclonal antibody [clone: 8.H.1]

Lieferant: US Biological
F0017
USBIF0017EA 418 EUR
USBIF0017
Anti-F9 Mouse monoclonal antibody [clone: 8.H.1]
Antikörper
Recommended dilution: ELISA: 5-10 ?g/ml; Immunochemistry: 100ng/ml

Type: Primary
Antigen: F9 (coagulation factor IX)
Clonality: monoclonal
Clone: 8.H.1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR