Anti-FABP4 Goat Polyclonal Antibody

Lieferant: US Biological
F0019-73A
USBIF0019-73AEA 555 EUR
USBIF0019-73A
Anti-FABP4 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FABP4 (fatty acid binding protein 4, adipocyte)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR