Anti-FABP4 Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
F0019-74E-BIOT
USBIF0019-74E-BIOTEA 884 EUR
USBIF0019-74E-BIOT
Anti-FABP4 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: FABP4 (fatty acid binding protein 4, adipocyte)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR