Anti-FABP4 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Lieferant: US Biological
F0019-74E-FITC
USBIF0019-74E-FITCEA 884 EUR
USBIF0019-74E-FITC
Anti-FABP4 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:50-1:100
Do not freeze!
Type: Primary
Antigen: FABP4 (fatty acid binding protein 4, adipocyte)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse