Anti-FABP4 Sheep Polyclonal Antibody

Lieferant: US Biological
F0019-74H
USBIF0019-74HEA 704 EUR
USBIF0019-74H
Anti-FABP4 Sheep Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FABP4 (fatty acid binding protein 4, adipocyte)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR