Anti-Fatty Acid Binding Protein, Heart Mouse Monoclonal Antibody (Biotin) [clone: 6A546]

Lieferant: US Biological
F0019-75J
USBIF0019-75JEA 520 EUR
USBIF0019-75J
Anti-Fatty Acid Binding Protein, Heart Mouse Monoclonal Antibody (Biotin) [clone: 6A546]
Antikörper
Recommended dilution: Western Blot: 1:50; Immunochemistry: 1:50

Type: Primary
Antigen: Fatty Acid Binding Protein, Heart
Clonality: monoclonal
Clone: 6A546
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Pig, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR