Anti-Fatty Acid Binding Protein, Liver Mouse Monoclonal Antibody (Biotin) [clone: 6A551]

Lieferant: US Biological
F0019-76P3
USBIF0019-76P3EA 520 EUR
USBIF0019-76P3
Anti-Fatty Acid Binding Protein, Liver Mouse Monoclonal Antibody (Biotin) [clone: 6A551]
Antikörper
Recommended dilution: Immunohistology: 1:10; Western Blot: 1:10

Type: Primary
Antigen: Fatty Acid Binding Protein, Liver
Clonality: monoclonal
Clone: 6A551
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR