Anti-Fetoprotein Mouse Monoclonal Antibody [clone: 10C232]

Lieferant: US Biological
F4100-01T
USBIF4100-01TEA 525 EUR
USBIF4100-01T
Anti-Fetoprotein Mouse Monoclonal Antibody [clone: 10C232]
Antikörper
Type: Primary
Antigen: Fetoprotein
Clonality: monoclonal
Clone: 10C232
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR