Anti-Fetoprotein Mouse Monoclonal Antibody [clone: D10]

Lieferant: US Biological
F4100-02E
USBIF4100-02EEA 843 EUR
USBIF4100-02E
Anti-Fetoprotein Mouse Monoclonal Antibody [clone: D10]
Antikörper
Type: Primary
Antigen: Fetoprotein
Clonality: monoclonal
Clone: D10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR