Anti-Fetoprotein Mouse Monoclonal Antibody [clone: 10g98]

Lieferant: US Biological
F4100-05S
USBIF4100-05SEA 608 EUR
USBIF4100-05S
Anti-Fetoprotein Mouse Monoclonal Antibody [clone: 10g98]
Antikörper
Type: Primary
Antigen: Fetoprotein
Clonality: monoclonal
Clone: 10g98
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR