Anti-Fibrin Degradation Product, D-Dimer Mouse Monoclonal Antibody [clone: 11C218]

Lieferant: US Biological
F4197-15P
USBIF4197-15PEA 879 EUR
USBIF4197-15P
Anti-Fibrin Degradation Product, D-Dimer Mouse Monoclonal Antibody [clone: 11C218]
Antikörper
Type: Primary
Antigen: Fibrin Degradation Product, D-Dimer
Clonality: monoclonal
Clone: 11C218
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR