Anti-Fibrin Degradation Product, D-Dimer Mouse Monoclonal Antibody [clone: 10B2317]

Lieferant: US Biological
F4197-15T
USBIF4197-15TEA 549 EUR
USBIF4197-15T
Anti-Fibrin Degradation Product, D-Dimer Mouse Monoclonal Antibody [clone: 10B2317]
Antikörper
Type: Primary
Antigen: Fibrin Degradation Product, D-Dimer
Clonality: monoclonal
Clone: 10B2317
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR