Anti-Fibrin Degradation Product, D-Dimer Mouse Monoclonal Antibody [clone: 9L169]

Lieferant: US Biological
F4197-17D
USBIF4197-17DEA 897 EUR
USBIF4197-17D
Anti-Fibrin Degradation Product, D-Dimer Mouse Monoclonal Antibody [clone: 9L169]
Antikörper
Type: Primary
Antigen: Fibrin Degradation Product, D-Dimer
Clonality: monoclonal
Clone: 9L169
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR