Anti-Fibrin Degradation Product, D-Dimer Mouse Monoclonal Antibody [clone: 9L170]
Lieferant: US Biological
F4197-17E
USBIF4197-17EEA 685 EUR
USBIF4197-17E
Anti-Fibrin Degradation Product, D-Dimer Mouse Monoclonal Antibody [clone: 9L170]
Antikörper
Type: Primary
Antigen: Fibrin Degradation Product, D-Dimer
Clonality: monoclonal
Clone: 9L170
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human