Anti-Fibrin Degradation Product, D-Dimer Mouse Monoclonal Antibody (Biotin) [clone: 13A105]

Lieferant: US Biological
F4197-20H
USBIF4197-20HEA 1050 EUR
USBIF4197-20H
Anti-Fibrin Degradation Product, D-Dimer Mouse Monoclonal Antibody (Biotin) [clone: 13A105]
Antikörper
Type: Primary
Antigen: Fibrin Degradation Product, D-Dimer
Clonality: monoclonal
Clone: 13A105
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR