Anti-FGFR2 Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
F4305-16G-BIOT
USBIF4305-16G-BIOTEA 884 EUR
USBIF4305-16G-BIOT
Anti-FGFR2 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: FGFR2 (fibroblast growth factor receptor 2)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR