Anti-4-Fluoro-7-Nitrobenzofurazan Rabbit Polyclonal Antibody

Lieferant: US Biological
F5102-05
USBIF5102-05EA 540 EUR
USBIF5102-05
Anti-4-Fluoro-7-Nitrobenzofurazan Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: 4-Fluoro-7-Nitrobenzofurazan
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR