Anti-Fluorescein Goat Polyclonal Antibody (TRITC (Rhodamine))

Lieferant: US Biological
F5200-30A
USBIF5200-30AEA 1150 EUR
USBIF5200-30A
Anti-Fluorescein Goat Polyclonal Antibody (TRITC (Rhodamine))
Antikörper
Type: Tag
Antigen: Fluorescein (fluorescein)
Clonality: polyclonal
Clone:
Conjugation: TRITC (Rhodamine)
Epitope:
Host: Goat
Isotype:
Reactivity: Goat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR