Anti-FGFR1OP Mouse Monoclonal Antibody [clone: 10K202]

Lieferant: US Biological
F5950-02
USBIF5950-02EA 540 EUR
USBIF5950-02
Anti-FGFR1OP Mouse Monoclonal Antibody [clone: 10K202]
Antikörper
Recommended dilution: Western Blot: 1:1000; Immunochemistry: 1:100; Immunoprecipitation: 1-2?g/?l

Type: Primary
Antigen: FGFR1OP (FGFR1 oncogene partner)
Clonality: monoclonal
Clone: 10K202
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR