Anti-Fructosyl Amino Acid Oxidase Sheep Polyclonal Antibody (Biotin)

Lieferant: US Biological
F8060-16B
USBIF8060-16BEA 843 EUR
USBIF8060-16B
Anti-Fructosyl Amino Acid Oxidase Sheep Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 1:8000-1:34000

Type: Primary
Antigen: Fructosyl Amino Acid Oxidase
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: E. coli
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR