Anti-FOXI1, pan Rabbit Polyclonal Antibody (KLH (Keyhole Limpet Hemocyanin))

Lieferant: US Biological
F9047-40-100
USBIF9047-40-100EA 270 EUR
USBIF9047-40-100
Anti-FOXI1, pan Rabbit Polyclonal Antibody (KLH (Keyhole Limpet Hemocyanin))
Antikörper
Recommended dilution: ELISA: 1: 200000; Western Blot: 1 ?g/ml

Type: Primary
Antigen: FOXI1, pan
Clonality: Polyclonal
Clone:
Conjugation: KLH (Keyhole Limpet Hemocyanin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR