Anti-FXYD Domain-containing Ion Transport Regulator 1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Lieferant: US Biological
F9400-04M-AP
USBIF9400-04M-APEA 677 EUR
USBIF9400-04M-AP
Anti-FXYD Domain-containing Ion Transport Regulator 1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500
Do not freeze!

Type: Primary
Antigen: FXYD Domain-containing Ion Transport Regulator 1
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR