Anti-FXYD Domain-containing Ion Transport Regulator 1 Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
F9400-04M-BIOT
USBIF9400-04M-BIOTEA 677 EUR
USBIF9400-04M-BIOT
Anti-FXYD Domain-containing Ion Transport Regulator 1 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: FXYD Domain-containing Ion Transport Regulator 1
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR