Anti-FXYD Domain-containing Ion Transport Regulator 1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Lieferant: US Biological
F9400-04M-HRP
USBIF9400-04M-HRPEA 677 EUR
USBIF9400-04M-HRP
Anti-FXYD Domain-containing Ion Transport Regulator 1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500
Type: Primary
Antigen: FXYD Domain-containing Ion Transport Regulator 1
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse