Anti-FH Goat Polyclonal Antibody

Lieferant: US Biological
F9508
USBIF9508EA 601 EUR
USBIF9508
Anti-FH Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:64000; Western Blot: 0. 01-0. 03?g/ml

Type: Primary
Antigen: FH (fumarate hydratase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR