Anti-MOGS Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Lieferant: US Biological
G2023-47A-APC
USBIG2023-47A-APCEA 884 EUR
USBIG2023-47A-APC
Anti-MOGS Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500
Do not freeze!
Type: Primary
Antigen: MOGS (mannosyl-oligosaccharide glucosidase)
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse