Anti-MOGS Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
G2023-47A-BIOT
USBIG2023-47A-BIOTEA 884 EUR
USBIG2023-47A-BIOT
Anti-MOGS Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: MOGS (mannosyl-oligosaccharide glucosidase)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR