Anti-GCP 2 Rabbit Polyclonal Antibody (Biotin)
Lieferant: US Biological
G2023-55A-50
USBIG2023-55A-50EA 608 EUR
USBIG2023-55A-50
Anti-GCP 2 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: GCP 2
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human