Anti-GDP-Mannose Pyrophosphorylase B, Isoform 2 Chicken polyclonal antibody

Lieferant: US Biological
G2023-92A
USBIG2023-92AEA 475 EUR
USBIG2023-92A
Anti-GDP-Mannose Pyrophosphorylase B, Isoform 2 Chicken polyclonal antibody
Antikörper
Recommended dilution: Western Blot: 1:1000-1:2000

Type: Primary
Antigen: GDP-Mannose Pyrophosphorylase B, Isoform 2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR