Anti-General Transcription Factor IIA, Polypeptide 1 Rabbit polyclonal antibody

Lieferant: US Biological
G2024-59
USBIG2024-59EA 768 EUR
USBIG2024-59
Anti-General Transcription Factor IIA, Polypeptide 1 Rabbit polyclonal antibody
Antikörper
Recommended dilution: Western Blot: 1:200-1:1000

Type: Primary
Antigen: General Transcription Factor IIA, Polypeptide 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR