Anti-GFAP Mouse monoclonal antibody [clone: GF-01]

Lieferant: US Biological
G2032-25J
USBIG2032-25JEA 633 EUR
USBIG2032-25J
Anti-GFAP Mouse monoclonal antibody [clone: GF-01]
Antikörper
Recommended dilution: Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: GFAP (glial fibrillary acidic protein)
Clonality: monoclonal
Clone: GF-01
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Pig
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR