Anti-GLA Rabbit Polyclonal Antibody

Lieferant: US Biological
G2035-48
USBIG2035-48EA 649 EUR
USBIG2035-48
Anti-GLA Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:1000; Immuncytochemistry/; Immunofluorescence: 1:10 -1:50; Immunochemistry: 1:10 -1:50

Type: Primary
Antigen: GLA (galactosidase, alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR