Anti-Gibberellic Acid Rat Polyclonal Antibody (Unconjugated)

Lieferant: US Biological
G2035-87
USBIG2035-87EA 795 EUR
USBIG2035-87
Anti-Gibberellic Acid Rat Polyclonal Antibody (Unconjugated)
Antikörper
Type: Primary
Antigen: Gibberellic Acid
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR