Anti-Glucosamine Rabbit Polyclonal Antibody

Lieferant: US Biological
G3040-89
USBIG3040-89EA 649 EUR
USBIG3040-89
Anti-Glucosamine Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100

Type: Primary
Antigen: Glucosamine
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR